Learn More
Scientific Industries SI™ Vortex-Genie™ 2T
Product Code. 15537365 Shop All Ohaus Products
Change view
Click to view available options
Plug Type:
EU Plug
Model:
SI-T256
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type Model Unit Size
15537365 EU Plug SI-T256 Each
10321432 EU Plug SI-T256 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product is under eco-contribution tax.
Product Code. 15537365 Supplier Scientific Industries SI™ Supplier No. 83041250
  • ONLINE EXCLUSIVE PRICE
    510.07€
    484.10€ / Pack of 1
    Save 25.97€
    5% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
Includes an integrated timer. Scientific Industries SI™ Vortex-Genie™ 2T mixes gently or vigorously with a set time or continuously.
  • Variable speed for precise mixing from gentle to vigorous
  • Broad range of attachments for most tubes, plates and other containers
  • Hands-Free or Touch On control
  • May be used in cold rooms and incubators
  • Timer function:
    • Touch On: 1 to 60 sec.
    • Hands-Free: 1 to 60 min. or continuous
  • Unit automatically stops at the end of user-programmed time
TRUSTED_SUSTAINABILITY
Specifications
Speed Range 600 to 2700 rpm
Electrical Requirements 230 V, 50 Hz
Voltage 230 V
Plug Type EU Plug
Frequency 50 Hz
Depth (Metric) 165 mm
Height (Metric) 165 mm
Width (Metric) 122 mm
Weight (Metric) 4 kg
Model SI-T256
Includes 76mm platform, pop-off cup