missing translation for 'onlineSavingsMsg'
Learn More

VP 14637, TRC

Product Code. 30455585
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30455585 100 mg 100mg
30455620 5 mg 5mg
30449889 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30455585 Supplier Toronto Research Chemicals Supplier No. TRCV785000100MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 235106-62-4
Molecular Formula C25 H22 N10 O3
Molecular Weight (g/mol) 510.51
Synonym 2,2'-[(4-Hydroxyphenyl)methylene]bis[4-[[(5-methyl-1H-tetrazol-1-yl)imino]methyl]phenol
IUPAC Name 2-[[2-hydroxy-5-[(E)-(5-methyltetrazol-1-yl)iminomethyl]phenyl]-(4-hydroxyphenyl)methyl]-4-[(E)-(5-methyltetrazol-1-yl)iminomethyl]phenol
SMILES Cc1nnnn1\N=C\c2ccc(O)c(c2)C(c3ccc(O)cc3)c4cc(\C=N\n5nnnc5C)ccc4O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C25H22N10O3/c1-15-28-30-32-34(15)26-13-17-3-9-23(37)21(11-17)25(19-5-7-20(36)8-6-19)22-12-18(4-10-24(22)38)14-27-35-16(2)29-31-33-35/h3-14,25,36-38H,1-2H3/b26-13+,27-14+
Formula Weight 510.1876
Chemical Name or Material VP 14637
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.