missing translation for 'onlineSavingsMsg'
Learn More

VP 14637, TRC

Product Code. 30455620
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455620 5 mg 5mg
30455585 100 mg 100mg
30449889 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30455620 Supplier Toronto Research Chemicals Supplier No. TRCV7850005MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 235106-62-4
Molecular Formula C25 H22 N10 O3
Molecular Weight (g/mol) 510.51
Synonym 2,2'-[(4-Hydroxyphenyl)methylene]bis[4-[[(5-methyl-1H-tetrazol-1-yl)imino]methyl]phenol
IUPAC Name 2-[[2-hydroxy-5-[(E)-(5-methyltetrazol-1-yl)iminomethyl]phenyl]-(4-hydroxyphenyl)methyl]-4-[(E)-(5-methyltetrazol-1-yl)iminomethyl]phenol
SMILES Cc1nnnn1\N=C\c2ccc(O)c(c2)C(c3ccc(O)cc3)c4cc(\C=N\n5nnnc5C)ccc4O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C25H22N10O3/c1-15-28-30-32-34(15)26-13-17-3-9-23(37)21(11-17)25(19-5-7-20(36)8-6-19)22-12-18(4-10-24(22)38)14-27-35-16(2)29-31-33-35/h3-14,25,36-38H,1-2H3/b26-13+,27-14+
Formula Weight 510.1876
Chemical Name or Material VP 14637
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.