missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ VP-SFM AGT™ Medium
VP-SFM AGT™ Medium
Brand: Gibco™ 12559003
Additional Details : Weight : 10.00000kg
Description
VP-SFM AGT™ is a serum-free, ultra-low protein medium in an easy-to-use granular format. It is designed for the growth of VERO cells for virus production and for the growth of COS-7, MDCK, BHK-21 and Hep-2 cells for recombinant protein production.Advanced Granulation Technology = AGT
Specifications
2 to 8°C | |
BHK-21, COS-7, HEp2, MDCK, VERO | |
AGT Medium | |
Phenol Red, Low Glucose, Sodium Pyruvate | |
Low | |
Antibody and cell culture production, bioproduction, mammalian cell culture, viral vaccine production | |
10 kg |
VP-SFM AGT™ Medium, 10kg, Powder Form, Ropak Bucket Format, Non-sterile, BHK-21, COS-7, HEp2, MDCK, VERO Cell Line, With Phenol Red, Low Glucose and Sodium Pyruvate, Without Glutamine and Antibiotics, Low Endotoxin Level, HEPES, Sodium Bicarbonate B | |
Powder (AGT) | |
Non-sterile | |
No Glutamine, No Antibiotics | |
Ropak bucket | |
Certified | |
For research use only |
For Research Use Only. Not for use in diagnostic procedures.