Learn More
VPS13C Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28676-1-AP-20UL
 View more versions of this product
Product Code. 16826215
  • 145.00€ / 20µL
Description
The VPS13C antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human VPS13C. This antibody recognizes human, mouse antigen. The VPS13C antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
VPS13C
Polyclonal
Unconjugated
20 μL
54832
IgG
Western Blot, ELISA
333 μg/mL
Rabbit
Primary
Human, Mouse