Learn More
VPS35 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10236-1-AP-150UL
Description
The VPS35 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human VPS35. This antibody recognizes human, mouse, rat antigen. The VPS35 antibody has been validated for the following applications: IF, IP, WB, ELISA analysis.
Specifications
| VPS35 | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 55737 | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, Co-immunoprecipitation (Co-IP) | |
| 260 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |