missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ VU 6015929

Description
VU 6015929 is a potent and selective discoidin domain receptor 1/2 (DDR1/2) inhibitor (IC50 = 4.67 nM and 7.39 nM for DDR1 and DDR2, respectively). VU 6015929 blocks collagen-induced DDR1 activation and collagen-IV production in HEK293 cells.
Specifications
Specifications
| Chemical Name or Material | 4-Fluoro-3-[[[5-(1-methyl-1 H-pyrazol-3-yl)-3-pyridinyl]methyl]amino]-N-[3-(trifluoromethoxy)phenyl]benzamide |
| CAS | 2442597-56-8 |
| Quantity | 10 mg |
| Target | DDR1/DDR2 |
| Molecular Formula | C24H19F4N5O2 |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?