Learn More
VWF Monoclonal Antibody (OTI8A6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803275S
Description
VWF Monoclonal antibody specifically detects VWF in Human samples. It is validated for Western BlotSpecifications
| VWF | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P04275 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7450 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI8A6 | |
| Unconjugated | |
| VWF | |
| 6820430P06Rik; AI551257; B130011O06Rik; C630030D09; Coagulation Factor VIII; coagulation factor VIII VWF; EMBL:CAB37852.1}; F8VWF; factor viii related antigen; von Willebrand antigen 2; von Willebrand antigen II; von Willebrand Disease (vWD); von Willebrand factor; Von Willebrand factor homolog; VWD; vwf; vwf {ECO:0000312 | |
| Human recombinant protein fragment corresponding to amino acids 764-1263 of human VWF produced in SF9 cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.