Learn More
Lovibond™ W600 Borosilicate Glasses
Specifications
Specifications
| Type | W600 Cuvette |
| Material | Borosilicate Glass |
| Quantity | 1Each |
| Path Length | 50 mm |
| For Use With (Equipment) | Model Fs, PFXi/PFX195, 880, 950 and 995 instruments |