Learn More
Heinz Herenz WA2 Wooden Applicator Single Cotton Bud Single End Tip
Product Code. 16328237
Change view
Click to view available options
Tip Type:
Single End
Unit Size:
Pack of 500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tip Type Unit Size
16328237 Single End Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16328237 Supplier Heinz Herenz Supplier No. 1030818
  • ONLINE EXCLUSIVE PRICE
    53.28€
    34.04€ / Pack of 500
    Save 19.24€
    36% Off
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
WA2 wooden applicator with single cotton bud.
We offer a wide range of sterile and nonsterile swabs:
  • Wood, PS, PP, aluminium or paper applicator
  • Each in various lengths available
  • With cotton, viscose, polyester or foam head
  • Diameter of the heads: small, medium or large
  • This guarantees that the right instrument is available for every special application.
  • For example, the cotton-tipped swabs are recommended for use by ENT specialists.
  • Our sterile cotton buds are all individually wrapped.
Please note in the medical field:
  • Only suitable for single use
  • Use by medical personnel only
  • This applicator size: single cotton bud is 10 x 35 mm mm and wooden stick is 200 x 3 mm
Specifications
Autoclavable Not Autoclavable
Sterility Non-sterile
Handle Material Wood
Product Type Cotton Swab
Tip Material Cotton
Tip Type Single End