Learn More
Esculape Laboratoire™ Wall First Aid Kits
Product Code. 11788373
Change view
Click to view available options
Height (Metric):
6 cm
9 cm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) unitSize
11788373 6 cm Each
11798373 9 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11788373 Supplier Esculape Laboratoire™ Supplier No. 716 01 05
  • ONLINE EXCLUSIVE PRICE
    39.90€
    21.63€ / Pack of 1
    Save 18.27€
    45% Off
Estimated Shipment: 13-08-2026
Specifications
Depth (Metric) 13 cm
Length (Metric) 18 cm
Product Type Wall First Aid Kit
Color White
Includes 1 Gas band 3m x 7cm, 1 Extensible web 3m x 7cm, 4 Alcoholic compresses, 2 Anti-compress shots, 2 Mosquito compress, 5 Sterile compresses 20 x 20, Guide first aid, 4 number of clips, scissors, Pair of Vinyl gloves, Sterile pressure dressing 2.5m x 10cm, st
Quantity 1 Ea.
Height (Metric) 6 cm