Learn More
Esculape Laboratoire™ Wall First Aid Kits
Product Code. 11798373
Change view
Click to view available options
Height (Metric):
6 cm
9 cm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
11798373 9 cm Each
11788373 6 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11798373 Supplier Esculape Laboratoire™ Supplier No. 719 01 02
  • ONLINE EXCLUSIVE PRICE
    75.60€
    41.91€ / Pack of 1
    Save 33.69€
    44% Off
Estimated Shipment: 18-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Depth (Metric) 20 cm
Length (Metric) 26 cm
Product Type Wall First Aid Kit
Color White
Includes 2 Gas band 3m x 7cm, 2 Extensible web 3m x 7cm, 3 Anti-compress shots, 10 Sterile compresses 20 x 20, Doigtier, Guide first aid, 6 number of clips, scissors, Pair of Vinyl gloves, Sterile pressure dressing 2.5m x 10cm, sterile pressure dressing 2.5 x 6cm,
Quantity 1 Ea.
Height (Metric) 9 cm