Learn More
Veolia Water Systems™ Wall-Mounted Demineralizer
Product Code. 11924877
Change view
Click to view available options
Flow Rate:
30 L/hr.
90 L/hr.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate Unit Size
11924877 30 L/hr. Each
11954887 90 L/hr. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11924877 Supplier Veolia Water Systems™ Supplier No. B114/A
  • ONLINE EXCLUSIVE PRICE
    1134.00€
    875.31€ / Pack of 1
    Save 258.69€
    22% Off
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.