Learn More
Hughes Wall-Mounted Eye and Face Shower with Open Basin
Product Code. 17712338
Change view
Click to view available options
Mounting:
Wall
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mounting Unit Size
17712338 Wall Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17712338 Supplier Hughes Supplier No. HEWD0085SS
  • ONLINE EXCLUSIVE PRICE
    609.29€
    319.51€ / Pack of 1
    Save 289.78€
    47% Off
Estimated Shipment: 03-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Wall mounted emergency eye/face wash with stainless steel open bowl.
  • Open stainless steel bowl, wall-mounted emergency eye/face wash featuring twin spray nozzles protected by dust covers which lift off upon activation via the push plate
  • The flow regulator valve guarantees an optimum water flow
  • The eye wash diffusers are fitted with fine mesh strainers and individual flow controllers designed to give a gentle flow of aerated water at 12 liters per minute as per the ANSI standard
  • This unit is intended for indoor use but can be located outdoors where there is no risk of overheating or freezing
Specifications
Depth (Metric) 30 cm
Material Stainless Steel
Mounting Wall
Product Type Emergency Shower
Width (Metric) 38 cm