Learn More
Haleco™ Waste Disposal Bags
Product Code. 12536239
Change view
Click to view available options
Capacity (Metric):
50 L
100 L
Unit Size:
Pack of 200
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12536239 100 L Pack of 200
12526239 50 L Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12536239 Supplier Haleco™ Supplier No. SACG100ST
Save Up to 60% on Liquid Handling
Use promo code 28028
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Color Gray
Disposable Single-use
For Use With (Application) Multi-purpose
Material LDPE
Weight (Metric) 14.3 kg
Product Type Garbage Bag
Capacity (Metric) 100 L
Length (Metric) 950 mm
Width (Metric) 820 mm
Thickness (Metric) 50 μm