Learn More
Rogo Sampaic™ Watch Glasses
Product Code. 11708094
Change view
Click to view available options
Capacity (Metric):
100 mL
1000 mL
150 mL
2 L
25 mL
250 mL
3 L
400 to 600 mL
50 mL
Unit Size:
Pack of 10
Pack of 20
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11708094 150 mL Pack of 20
11702724 100 mL Pack of 20
11792714 25 mL Pack of 20
11738094 1000 mL Pack of 10
11728094 400 to 600 mL Pack of 20
11748094 2 L Pack of 10
11724146 3 L Pack of 10
11778084 50 mL Pack of 20
11718094 250 mL Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 11708094 Supplier Rogo Sampaic™ Supplier No. KBN014
Only null left
Add to basket
Edge
Add to basket
Excellent for liquid evaporation, weighing solids, heating small amounts of substances and as beaker covers. Rogo Sampaic™ Watch Glasses are manufactured from durable clear borosilicate glass.
  • Available in a range of diameters
  • Corrosion-free and resistant to thermal shock
Specifications
Capacity (Metric) 150 mL
Material Borosilicate Glass
Type Watch Glass
For Use With (Application) Beaker
Temperature Range (Metric) 500 °C
Diameter (Metric) 70 mm