missing translation for 'onlineSavingsMsg'
Learn More

Water, Molecular Biology Grade, Sterile, Ricca Chemical

Product Code. 31751156
Change view
Click to view available options
Quantity:
1 L
Packaging:
Clear PET-G Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
31751156 1 L Clear PET-G Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31751156 Supplier Ricca Chemical Company Supplier No. R91450001G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

RNase Free, DNase Free, Protease Free, DEPC Free

  • Molecular biology grade water is free of Rnase, Dnase, Protease enzymes
  • DEPC free
  • Ideal for many fundamental procedures such as PCR, electrophoresis, DNA sequencing, and buffers for enzymatic analyses

Specifications

pH 5 to 7
CAS 7732-18-5
Color Clear, Colorless
Melting Point 0°C
Boiling Point 100°C
DNase No
Molecular Formula H2O
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
Solubility Information Miscible
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name oxidane
Molecular Weight (g/mol) 18.015
PubChem CID 962
ChEBI CHEBI:15377
Formula Weight 18.01
Percent Purity 100%
Grade Molecular Biology
Odor Odorless
Packaging Clear PET-G Bottle
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Water, Sterile, Dnase, Rnase, Protease, DEPC free
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.