missing translation for 'onlineSavingsMsg'
Learn More

Water Pollution QC Check Sample: Total Phenolics

Product Code. 30325465
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30325465 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325465 Supplier VHG Labs Supplier No. VHGQWPPHEN2
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenolics
Analyte or Component Names Phenolics
Shipping Condition Room Temperature
Concentration 0.775 mg/L
Synonym Phenolics
Solution Type Mixture
IUPAC Name Phenolics
Solvent or Matrix Methanol
Industry Type Petrochemical Testing
Quantity 2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.