Learn More
Thermo Scientific™ Tempshield™ Cryo Gloves - Elbow Length
Product Code. p-7219196 Shop All Thermo Scientific Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
Unit Size:
1 pair
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
16429019 Medium 1 pair
15342547 Large 1 pair
16439019 X-Large 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16429019 Supplier Thermo Scientific™ Supplier No. 189445
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Multi-layered construction allows for thermal protection, flexibility, and dexterity which are essential features when function is important and safety is critical. High performance, thermal inner lining wicks moisture away from hands, maintaining comfort over extended periods.
Multi-layered construction allows for thermal protection, flexibility, and dexterity which are essential features when function is important and safety is critical. High performance, thermal inner lining wicks moisture away from hands, maintaining comfort over extended periods. Safety certified to meet EU standards for CE Cat. III: EN511, EN388, EN ISO 21420. Available in wrist, elbow, mid-arm, shoulder-length options.
  • Provides splash protection
  • Comfortable, lightweight, breathable
Certifications
CE Cat. III: EN 511, EN 388, EN ISO 21420
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE Cat. III: EN 511, EN 388, EN ISO 21420
Description Elbow Length
Product Type Waterproof Cryogenic Gloves
Size Medium
For Use With (Application) Cryogenic Preservation
Length (English) 18 in.
Length (Metric) 45.7 cm
CE Cat. III: EN 511, EN 388, EN ISO 21420