missing translation for 'onlineSavingsMsg'
Learn More

Waters - Trace Elements Spiking Solution EG1: Ag, Be, Cd @ 5; Co @ 20; Cr, Cu, Mn, Mo, Ni, Pb, Sb, Tl, V @ 40; Al, Fe, Se, Zn @ 100; As, Ba, SiO2 @ 200 μg/mL in 5% HNO3, tr. HF, tr. Tartaric Acid, VHG™

Product Code. 30002945
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30002945 100 mL 100mL
17948029 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30002945 Supplier VHG Labs Supplier No. VHGSSEG1100
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Waters - Trace Elements Spiking Solution EG1: Ag, Be, Cd @ 5; Co @ 20; Cr, Cu, Mn, Mo, Ni, Pb, Sb, Tl, V @ 40; Al, Fe, Se, Zn @ 100; As, Ba, SiO2 @ 200 μg/mL in 5% HNO3, tr. HF, tr. Tartaric Acid
TRUSTED_SUSTAINABILITY

Spezifikation

Chemical Name or Material Beryllium/Cadmium/Silver/Cobalt/Antimony/Chromium/Copper/Lead/Manganese/Molybdenum/Nickel/Thallium/Vanadium/Aluminium/Iron/Selenium/Arsenic/Barium/Silicon dioxide/Zinc
CAS 7440-41-7, 7440-43-9, 7440-22-4, 7440-48-4, 7440-36-0, 7440-47-3
Methods ICP/ICP-MS
Analyte or Component Names Beryllium/Cadmium/Silver/Cobalt/Antimony/Chromium/Copper/Lead/Manganese/Molybdenum/Nickel/Thallium/Vanadium/Aluminium/Iron/Selenium/Arsenic/Barium/Silicon dioxide/Zinc
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dilute HNO3
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Beryllium 5 μg/mL/Cadmium 5 μg/mL/Silver 5 μg/mL/Cobalt 20 μg/mL/Antimony 40 μg/mL/Chromium 40 μg/mL/Copper 40 μg/mL/Lead 40 μg/mL/Manganese 40 μg/mL/Molybdenum 40 μg/mL/Nickel 40 μg/mL/Thallium 40 μg/mL/Vanadium 40 μg/mL/Aluminium 100 μg/mL/Iron 100 μg/mL/Selenium 100 μg/mL/Arsenic 200 μg/mL/Barium 200 μg/mL/Silicon dioxide 200 μg/mL/Zinc 1000 μg/mL
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.