missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDFY4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17558-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody detects the C-terminal of WDFY4. The gene WDFY4 has several isoforms: 354 kDa, 73 kDa, 115 kDa, 353 kDa, 325 kDa, 222 kDa.
WDFY4, also named as C10orf64 and KIAA1607, is a huge protein with unknown functions. WDFY4 is expressed predomitly in immune tissues such as lymph nodes, the spleen, the thymus and the tonsils. According to recent genome-wide association studies (GWAS), WDFY4 was introduced as novel predisposing genes for SLE (Systemic Lupus Erythematosus) which is a heterogeneous complex relapsing-remitting autoimmune disease.Specifications
| WDFY4 | |
| Polyclonal | |
| Unconjugated | |
| WDFY4 | |
| C10orf64, KIAA1607, WDFY family member 4, WDFY4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 498582, 545030, 57705 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6ZS81 | |
| Wdfy4 | |
| WDFY4 Fusion Protein Ag11720 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction