Learn More
WDR19 Mouse anti-Human, Unconjugated, Clone: 2D1H4, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | WDR19 |
|---|---|
| Clone | 2D1H4 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16809905
|
Proteintech
67330-1-IG-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16899895
|
Proteintech
67330-1-IG-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The WDR19 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human WDR19. This antibody recognizes human antigen. The WDR19 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| WDR19 | |
| 1000 μg/mL | |
| Monoclonal | |
| Mouse | |
| 57728 | |
| Primary |
| 2D1H4 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Human | |
| IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.