missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR62 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-77302H
511.20 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
WDR62 Polyclonal antibody specifically detects WDR62 in Human, Mouse samples. It is validated for Western Blot
Specifications
WDR62 | |
Polyclonal | |
Western Blot | |
C19orf14MGC166976, chromosome 19 open reading frame 14, DKFZP434J046, DKFZp686G1024, EC 2.7.11.1, EC 2.7.7.38, FLJ33298, WD repeat domain 62, WD repeat-containing protein 62 | |
The immunogen recognized by this antibody maps to a region between residue 750 and 800 of human WD repeat domain 62 using the numbering given in entry NP_775907.4 (GeneID 284403). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
HRP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
284403 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction