Learn More
WDR92 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25540-1-AP-20UL
 View more versions of this product
Product Code. 16884105
  • 145.00€ / 20µL
Description
WD40 repeats are a common structural module in eukaryotic proteins, and proteins containing WD40 domains have a wide range of functions, including signal transduction, cell cycle regulation, RNA splicing, and transcription. One such protein, WDR92, also known as monad, contains two WD40 repeats is widely expressed in human tissues, especially testis. Overexpression of this protein or its binding partner RNA polymerase II-associated protein 3 (RPAP3) potentiated apoptosis and caspase-3 activation induced by TNF-a and cycloheximide, suggesting that WDR92, together with RPAP3, may function as a novel modulator of apoptosis.
Specifications
Specifications
WDR92
Polyclonal
Unconjugated
WDR92
WD repeat domain 92, WDR92
Rabbit
Antigen Affinity Chromatography
RUO
103784, 116143
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence
0.16 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8BGF3, Q96MX6
Wdr92
WDR92 Fusion Protein Ag22181
20 μL
Primary
Mouse, Human