Learn More
Kartell SpA Weighing Bottles
Description
Airtight push on lids included. Highly resistant to acid attack. No internal shoulders to allow use of spatulas. Tops can be squeezed to assist pouring. Remove lid before autoclaving.
Specifications
Specifications
| Capacity (Metric) | 23 mL |
| Color | Clear |
| Material | Polypropylene |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Fit for Purpose | Certified Quality |
| Autoclavable | Autoclavable |
| Capacity (English) | 0.78 oz. |
| Shape | Round |
| Disposable | Reusable |
| Show More |