missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WFDC2 Recombinant Rabbit Monoclonal Antibody (17D2)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA560684

Product Code. 30533732

  • 459.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

WFDC2 Recombinant Monoclonal Antibody for Western Blot, Flow, ELISA

This gene encodes a protein that is a member of the WFDC domain family. The WFDC domain, or WAP Signature motif, contains eight cysteines forming four disulfide bonds at the core of the protein, and functions as a protease inhibitor in many family members. This gene is expressed in pulmonary epithelial cells, and was also found to be expressed in some ovarian cancers. The encoded protein is a small secretory protein, which may be involved in sperm maturation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

WFDC2
Recombinant Monoclonal
1.5 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q14508
WFDC2
A synthesized peptide derived from Human WFDC2.
100 μL
Primary
Human
Antibody
IgG
ELISA, Flow Cytometry, Western Blot
17D2
Unconjugated
WFDC2
1600023A02Rik; dJ461P17.6; EDDM4; epididymal protein 4; epididymal secretory protein 4; Epididymal secretory protein E4; epididymis-specific, whey-acidic protein type, four-disulfide core; HE4; Major epididymis-specific protein E4; MGC57529; putative protease inhibitor WAP5; RE4; WAP domain containing protein HE4-V4; WAP domain-containing protein HE4; WAP four-disulfide core domain 2; WAP four-disulfide core domain protein 2; WAP5; Wfdc2
Rabbit
Affinity chromatography
RUO
10406
-20°C or -80°C if preferred
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.