missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GE Healthcare Whatman™ Flexible-Backed TLC Plates
Description
- 20 × 20cm size
- Adsorption plates have 60Å silica gel medium and 250μm layer thickness
- Ion-exchange plates (DEAE medium) have 100μm layer thickness and come without fluorescent indicator
Specifications
Specifications
| Fluorescent Indicator | Yes |
| For Use With (Application) | For mobile phases containing strong acids/bases |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?