Learn More
Cytiva Whatman™ Flexible Capmats
Specifications
Specifications
| Material | Silicone |
| Autoclavable | Autoclavable |
| No. of Wells | 96 |
| No. per Pack | 50 Pieces |
| Product Type | Cap Mat |
| Well Shape | Round |
| Plate Compatibility | 96-well Microplate, 300, 750 and 800 μL |