Learn More
GE Healthcare Whatman™ Klari-Flex™ Bottle-top Filtration System
Product Code. 11958149
Change view
Click to view available options
:
Pore size: 0.22μm PES; Funnel and bottle: 500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11958149 Pore size: 0.22μm PES; Funnel and bottle: 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11958149 Supplier GE Healthcare Supplier No. 65145002
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Provide vacuum filtration of aqueous solutions such as cell culture media, biological fluids, and buffers
  • Drop-Connect technology ensures fail-safe vacuum connection and unit placement in base.
  • Weighted pedestal base and wide, low-profile funnel/bottle filtration units are stable and tip resistant, minimizing the risk of spills and contamination.
  • Graduation marks face front for readability.
  • Large bottle cap accommodates labeling information.
  • Ergonomically designed bottle and bottle cap provide improved handling.
  • Membrane material is PES.
TRUSTED_SUSTAINABILITY
Specifications
Description Funnel and bottle
Volume (Metric) 500 mL