Learn More
Cytiva Whatman™ Litmus Blue Test Paper
Description
Impregnated with a mixture of pH indicators, acid-alkali litmus papers assume a color specific to sample pH upon contact with a sample. Whatman™ Litmus Blue Test Paper provides rapid determination of pH values and has a variety of applications in laboratories and industry.
Mixed-indicator tests designed for fast, accurate, and reliable pH solution testing.
- Instant pH readings
- Accurate for a wide range of routine pH testing
- Convenient and portable for field use
- Color change in pH solutions ranging from approximately 5 to 8
- Economical
Specifications
Specifications
| Color Indicator | Blue |
| pH Range | 5 to 8 |
| Format | Reel |
| Quantity | 1 Each |
| Type | Test Paper |