Learn More
Cytiva Whatman™ pH Indicators Integral Comparison Strips
Product Code. 11370684
Change view
Click to view available options
pH Range:
1 to 12
1.8 to 3.8
3.8 to 5.5
5.2 to 6.8
6 to 8.1
8 to 9.7
9.5 to 12
Unit Size:
Pack of 200
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range unitSize
11370684 1 to 12 Pack of 200
10451602 3.8 to 5.5 Pack of 200
10114071 1.8 to 3.8 Pack of 200
10614282 5.2 to 6.8 Pack of 200
11330694 6 to 8.1 Pack of 200
11340694 8 to 9.7 Pack of 200
11914556 9.5 to 12 Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11370684 Supplier Cytiva Supplier No. 2612990
Only null left
Add to basket
Add to basket
Used for rapid determination of pH values
  • Instant pH readings
  • Accurate for wide range of routine pH testing
  • Convenient and portable for field use
TRUSTED_SUSTAINABILITY
Specifications
pH Range 1 to 12
Format Strip
For Use With (Application) Routine pH testing
Includes Plastic supports, paper segments on each strip, complementary indicator charts
Type pH Indicator Test Strip, Integral Comparison Strip
Dimensions (L x W) 100 x 11 mm