Learn More
Cytiva Whatman™ pH Indicators Integral Comparison Strips
Description
Used for rapid determination of pH values
- Instant pH readings
- Accurate for wide range of routine pH testing
- Convenient and portable for field use
Specifications
Specifications
| pH Range | 9.5 to 12 |
| Format | Strip |
| For Use With (Application) | Routine pH testing |
| Includes | Integral color chart |
| Type | pH Indicator Test Strip, Integral Comparison |
| Dimensions (L x W) | 100 x 11 mm |