Learn More
Qiagen SPOT CloneSaver Holder
Description
SPOT CloneSaver Holder is a rigid frame that allows automated spotting to standard CloneSaver Cards. It keeps the card flat for uniform and precise spotting of biological samples. The 96-well card is easily inserted into the ANSI/SBS-compatible frame, which can then be placed onto a liquid-handling deck just like a multiwell plate.
The SPOT CloneSaver Holder is compatible with standard liquid handlers manufactured by companies such as Beckman Coulter and Tecan Instruments.
The SPOT CloneSaver Holder is compatible with standard liquid handlers manufactured by companies such as Beckman Coulter and Tecan Instruments.
Specifications
Specifications
| Product Type | CloneSaver Card Holder |
| Description | Keeps card flat for uniform, precise spotting of biological samples |
| For Use With (Equipment) | Compatible w/standard liquid handlers |