Learn More
Cytiva Whatman™ Stopper for use with MV050 Vacuum Filtration Manifolds
Description
- Product Type: Stopper
- Material: Rubber
- For Use with (Equipment): MV050 Vacuum Filtration Manifolds
- For Use with (Applications): Microbiology, Biochemistry, and Hydrobiology. Residue Analysis, Precipitate Analysis, and Contamination Testing (Food and Beverage, Pharmaceutical, and Cosmetic Industries)
Specifications
Specifications
| For Use With (Application) | Microbiology, Biochemistry, and Hydrobiology. Residue Analysis, Precipitate Analysis, and Contamination Testing (Food and Beverage, Pharmaceutical, and Cosmetic Industries) |
| For Use With (Equipment) | MV050 Vacuum Filtration Manifolds |
| Autoclavable | Autoclavable |
| Material | Rubber |
| Product Type | Stopper (1 hole) |
| Sterility | Non-sterile |