Learn More
DWK Life Sciences WHEATON™ CompletePAK™ Sterile Components Stoppers
Product Code. 17932611 Shop All Wheaton Science Products Products
Change view
Click to view available options
Closure Size:
13 mm
20 mm
Style:
OmniFlex 3G Igloo
OmniFlex 3G Straight Plug
Ultra Pure Straight Plug
Unit Size:
Pack of 125
Pack of 285
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Size Style Unit Size
17932611 13 mm OmniFlex 3G Straight Plug Pack of 285
17952611 13 mm OmniFlex 3G Igloo Pack of 285
17902581 13 mm Ultra Pure Straight Plug Pack of 285
17952621 20 mm OmniFlex 3G Igloo Pack of 125
17962621 20 mm OmniFlex 3G Straight Plug Pack of 125
17962611 20 mm Ultra Pure Straight Plug Pack of 125
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17932611 Supplier DWK Life Sciences Supplier No. W988003S
  • ONLINE EXCLUSIVE PRICE
    1243.00€
    1191.72€ / Pack of 285
    Save 51.28€
    4% Off
Estimated Shipment: 02-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.
  • Sterile
  • Stopper
Certifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
Specifications
Sterility Sterile
No. per Case 285/Cs.
Style OmniFlex 3G Straight Plug
Product Line Wheaton CompletePAK
Closure Size 13 mm
Certifications/Compliance USP 788, 85, and 71, Container Closure Integrity (CCI) certified