Learn More
DWK Life Sciences Wheaton™ Crimpers
Product Code. 10670884 Shop All Wheaton Science Products Products
Change view
Click to view available options
For Use With (Equipment):
11 mm Seals
13 mm Seals
20 mm Seals
8 mm Seals
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10670884 11 mm Seals Each
11591215 8 mm Seals Each
10495951 13 mm Seals Each
10432722 20 mm Seals Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10670884 Supplier DWK Life Sciences Supplier No. W225301
  • ONLINE EXCLUSIVE PRICE
    558.60€
    552.64€ / Pack of 1
    Save 5.96€
    1% Off
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
With ergonomically designed handles for sealing standard laboratory vials
  • Wheaton E-Z Crimpers feature cushioned ergonomic handles that aid in reducing hand fatigue and add a high degree of comfort for the user. Each crimper size is labeled for quick identification. Other features include polished crimping jaws for consistent sealing, a short handle and redesigned spring assembly that further aids in reducing hand fatigue. For semi-automatic crimping, use the E-Z Crimper with the Wheaton Crimpmaster™ Crimping Station. An adjustable stop allows you to determine the tightness of the seal while maintaining consistent crimping. Tapered jaws provide for easier vial access when using racks. The 11 mm E- Z Crimper is supplied with a plunger insert to give the option of crimping the top of a vial creating an extra tight seal. We recommend this only for standard opening vials.
  • Crimpers can be autoclaved, although autoclaving may change the appearance or reduce product life. They should not be autoclaved at greater than 121°C
Warranty
Two years
Specifications
Compatible Storage Tubes Tubes with 11 mm Crimp Caps
Operation Manual
Type Manual Crimper
For Use With (Equipment) 11 mm Seals
Size 11 mm
Quantity 1 Ea.