Learn More
DWK Life Sciences Wheaton™ Reusable PVC Coliwasas and Accessories
Product Code. 11734129 Shop All Wheaton Science Products Products
Change view
Click to view available options
Product Type:
Coliwasa Sampler
Extension and Connector
Length (Metric):
1.21 m
1.82 m
Length (English):
4 ft.
6 ft.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Length (Metric) Length (English) unitSize
11734129 Extension and Connector 1.21 m 4 ft. Each
12437311 Coliwasa Sampler 1.82 m 6 ft. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11734129 Supplier DWK Life Sciences Supplier No. 885253
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
For a variety of applications
TRUSTED_SUSTAINABILITY
Specifications
Product Type Extension and Connector
Capacity (English) 25.6 oz.
Capacity (Metric) 760 mL
For Use With (Equipment) Clear PVC reusable coliwasas
Length (English) 4 ft.
Length (Metric) 1.21 m