Learn More
DWK Life Sciences Wheaton™ Flat Bottom Flasks For Soxhlet Extraction Apparatus
Product Code. 12457111 Shop All Wheaton Science Products Products
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
12457111 250 mL Each
12447111 125 mL Each
12467111 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12457111 Supplier DWK Life Sciences Supplier No. 428146
Trusted Glassware Deals
Use promo code 28509
View all promotions
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
Manufactured from WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements
  • Flat bottom flask 24/41
TRUSTED_SUSTAINABILITY
Specifications
Description 1/case
Capacity (Metric) 250 mL
Material Borosilicate Glass
For Use With (Application) For continuous extraction of analytes from a solid into an organic solvent
Standard Taper Joint 24/40
Bottom Shape Flat