Learn More
Fisherbrand™ White Plastic General Purpose Trays
Product Code. 12397237 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
Dimensions (L x W):
460 x 260mm
570 x 340mm
570 x 540mm
680 x 540mm
700 x 460mm
1130 x 540mm
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
12397237 1130 x 540mm Each
12347237 460 x 260mm Each
12357237 570 x 340mm Each
12367237 570 x 540mm Each
12377237 680 x 540mm Each
12387237 700 x 460mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 12397237
  • ONLINE EXCLUSIVE PRICE
    117.00€
    114.62€ / Pack of 1
    Save 2.38€
    2% Off
Estimated Shipment: 16-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Available in general purpose, biohazard and radiation safety formats, spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection.
  • Available in general purpose, biohazard and radiation safety formats
  • Spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection
  • All trays supplied with one free liner when purchased
  • Additional environmentally friendly APET liners are provided in packs of 25
Specifications
Dimensions (L x W) 1130 x 540mm
Color White
Length (Metric) 1130 mm
Material Plastic
Type Tray
Width (Metric) 540 mm
For Use With Safe containment of spillages, lab bench protection
Includes Liner
No. per Pack 1 Each
Shape Rectangular