Learn More
TFA Dostmann Wind Meter WIND BEE
Product Code. 17931642
Change view
Click to view available options
Case Material:
Plastic
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Case Material Unit Size
17931642 Plastic Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 17931642 Supplier TFA Dostmann Supplier No. 42.6003.13
  • ONLINE EXCLUSIVE PRICE
    72.58€
    53.29€ / Pack of 1
    Save 19.29€
    26% Off
Estimated Shipment: 16-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
The handy wind meter WIND BEE displays the current wind speed and the highest and lowest values in kilometers or miles per hour, in meters per second or in knots. In addition, the temperature is determined. WIND BEE has a tripod connector for measurements over a longer period of time.
The handy anemometer WIND BEE by TFA is the ideal tool for all sailors, kite surfers and outdoor enthusiasts. WIND BEE is also a useful companion at work or in various leisure activities. The device displays the current wind speed and the highest and lowest values in kilometers or miles per hour, in meters per second or in knots. In addition, the temperature is determined. WIND BEE has a tripod connector for measurements over a longer period of time.
  • Ideal for easy measurement of wind speed (at work or during various leisure activities)
  • Highest and lowest values and display of the temperature
  • Accuracy ± 3%
  • Backlight
  • With tripod connector
Specifications
Type Anemometer
Accuracy ±3%
Case Material Plastic