Learn More
Branson Ultrasonics™ Wire Basket For Washer
Brand: Branson Ultrasonics™ 000-019-229
Additional Details : Weight : 0.27000kg
Specifications
1 Each | |
109 mm | |
88 mm | |
209 mm |
Wire Basket | |
Ultrasonic Washer, 2510/2800 Branson | |
Stainless Steel | |
2510/2800 |