missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ECE-1 Antibody (OTI2B6), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70612G
This item is not returnable.
View return policy
Description
ECE-1 Monoclonal specifically detects ECE-1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| ECE-1 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ECE1 | |
| Recombinant protein fragment corresponding to amino acids 486-770 of human ECE-1 (NP_001388) produced in E.coli. | |
| 0.1 mL | |
| 1889.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI2B6 | |
| Western Blot | |
| EC 3.4.24, EC 3.4.24.71, ECE, ECE-1, endothelin converting enzyme 1, endothelin-converting enzyme 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction