missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wnt-1 Antibody (10C8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-51575-0.025ml
This item is not returnable.
View return policy
Description
Wnt-1 Monoclonal specifically detects Wnt-1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Wnt-1 | |
| Monoclonal | |
| Unconjugated | |
| INT1Proto-oncogene Int-1 homolog, proto-oncogene Wnt-1, wingless-type MMTV integration site family, member 1, wingless-type MMTV integration site family, member 1 (oncogene INT1) | |
| Mouse | |
| 41 kDa | |
| 0.025 mL | |
| Cellular Markers, Oncogenes, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
| 7471 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| 10C8 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| WNT1 | |
| Purified recombinant fragment of WNT1 expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction