missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wnt-5a Antibody (CL13683) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24567MFV500
This item is not returnable.
View return policy
Description
Wnt-5a Monoclonal antibody specifically detects Wnt-5a in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| Wnt-5a | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| hWNT5A, protein Wnt-5a, wingless-type MMTV integration site family, member 5A, WNT-5A protein | |
| This antibody was developed using a synthetic peptide derived from P41221, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer Stem Cells, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
| 7474 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL13683 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction