missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wnt-9a Antibody (326711) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3157JF585
This item is not returnable.
View return policy
Description
Wnt-9a Monoclonal antibody specifically detects Wnt-9a in Human samples. It is validated for Western Blot,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| Wnt-9a | |
| Monoclonal | |
| Janelia Fluor 585 | |
| MGC138165, MGC141991, Protein Wnt-14, wingless-type MMTV integration site family, member 14, wingless-type MMTV integration site family, member 9A, WNT14protein Wnt-9a | |
| E. coli-derived recombinant human Wnt-9a, Pro48-Glu92 & Arg231-Phe304, Accession # O14904.2 | |
| 0.1 mL | |
| Signal Transduction, Wnt Signaling Pathway | |
| 7483 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 326711 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction