Learn More
Gilac™ Women's 1 L Urinal
Product Code. 17918981
Change view
Click to view available options
Color:
Green
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17918981 Green Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17918981 Supplier Gilac™ Supplier No. G123708
  • ONLINE EXCLUSIVE PRICE
    14.37€
    9.49€ / Pack of 1
    Save 4.88€
    33% Off
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Women's 1 L Urinal is is suitable for hospital use.
  • Suitable for hospital use.
  • Translucent, level visible, with volume graduations.
  • Rounded design and very thick.
  • Not easy to break when dropped.
  • Outstanding durability even in the intensive use cycle.
  • Product is disinfected in a bedpan washer.
  • Researched product ergonomics ensure optimum cleaning.
  • Equipment at room temperature to avoid the cold sensation experienced with stainless steel equivalents.
Specifications
Product Type Urinal
Color Green
Material Polypropylene
Sterility Non-sterile
Disposable Reusable
Graduated Yes
Volume (English) 0.3 gal.
Volume (Metric) 1 L