Learn More
Techsit Worktech Laboratory ESD Low Chair
Product Code. 30306781
Encompass_Preferred
Change view
Click to view available options
Style:
Castors
High back
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Style Unit Size
30306781 High back Each
30306779 Castors Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30306781 Supplier Techsit Supplier No. TS1920HB
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Low ESD vinyl Cleanroom chair on ESD
  • ESD Cleanroom vinyl low chair on ESD Castors
  • Black ESD/Anti-static Vinyl that is easy to clean
  • Fully adjustable Seat height, back height, and back rake
  • Ergonomically designed and contoured seat and back
  • Cleanroom class 7 suitable
  • BS EN61340-5-1
  • Conductivity 104 - 106 Ohms
Low ESD Vinyl Cleanroom Chair
Specifications
Product Type Cleanroom ESD Chair
Color Black
Autoclavable Not Autoclavable
Back Style Ergonomic
Style High back
With Armrest No
With Backrest Yes
With Casters Yes
Footrest No
With Lumbar Support Yes
With Tilt No
Material Vinyl Upholstery
Seating Type Desk Height