missing translation for 'onlineSavingsMsg'
Learn More

WP 155, TRC

Product Code. 30137895
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137895 1 g 1g
30003833 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137895 Supplier Toronto Research Chemicals Supplier No. TRCW4995501G
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1031-47-6
Molecular Formula C12 H19 N6 O P
Molecular Weight (g/mol) 294.29
Synonym P-(5-Amino-3-phenyl-1H-1,2,4-triazol-1-yl)-N,N,N',N'-tetramethylphosphonic diamide, 5-Amino-1-[bis(dimethylamino)phosphinyl]-3-phenyl-1,2,4-triazole, 5-Amino-1-bis(dimethylamido)phosphoryl-3-phenyl-1,2,4-triazole, 5-Amino-3-phenyl-1,2,4-triazol-1-yl-N,N,N',N'-tetramethylphosphodiamide, NSC 232670, Triamiphos, WP 155, Wepsyn, Wepsyn 155
IUPAC Name 2-[bis(dimethylamino)phosphoryl]-5-phenyl-1,2,4-triazol-3-amine
SMILES CN(C)P(=O)(N(C)C)n1nc(nc1N)c2ccccc2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H19N6OP/c1-16(2)20(19,17(3)4)18-12(13)14-11(15-18)10-8-6-5-7-9-10/h5-9H,1-4H3,(H2,13,14,15)
Formula Weight 294.1358
Percent Purity >95%
Chemical Name or Material Triamiphos
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.