Learn More
Melford X-GlcA Cyclohexylammonium Salt
Product Code. 30018105
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
1 g
Unit Size:
100mg
1g
250mg
500mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018105 500 mg 500mg
30018106 1 g 1g
30018103 100 mg 100mg
30018104 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30018105 Supplier Melford Supplier No. B722000.5
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
Histochemical substrate for β-D-Glucuronidase (GUS) encoded by the gusA gene.
Histochemical substrate for β-D-Glucuronidase (GUS) encoded by the gusA gene. The substrate is used as a qualitative histochemical marker of specific GUS expression in cells and tissue. X-GlcA is cleaved by GUS at the B1 glucuronic bond between glucuronic acid and the 5-Bromo-4-chloro-3-indolyl part of X-GlcA via hydrolysis. The cleavage of X-GlcA results in the precipitation of water insoluble blue dichloro-dibromo-indigo precipitate at the site of enzymatic cleavage. Allow to warm to room temperature before using.
Specifications
Quantity 500 mg