missing translation for 'onlineSavingsMsg'
Learn More

Melford X-GlcA Cyclohexylammonium Salt

Product Code. 30018106
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
1 g
Unit Size:
100mg
1g
250mg
500mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018106 1 g 1g
30018105 500 mg 500mg
30018103 100 mg 100mg
30018104 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30018106 Supplier Melford Supplier No. B722001.0
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Histochemical substrate for β-D-Glucuronidase (GUS) encoded by the gusA gene.

Histochemical substrate for β-D-Glucuronidase (GUS) encoded by the gusA gene. The substrate is used as a qualitative histochemical marker of specific GUS expression in cells and tissue. X-GlcA is cleaved by GUS at the B1 glucuronic bond between glucuronic acid and the 5-Bromo-4-chloro-3-indolyl part of X-GlcA via hydrolysis. The cleavage of X-GlcA results in the precipitation of water insoluble blue dichloro-dibromo-indigo precipitate at the site of enzymatic cleavage. Allow to warm to room temperature before using.

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.